missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perforin Mouse anti-Human, PE, Clone: dG9 (delta G9), eBioscience™

Mouse Monoclonal Antibody

$174.24 - $376.20

Specifications

Antigen Perforin
Clone dG9 (delta G9)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011353 Life Technologies
LS12999441
25 Tests Each for $174.24
5011354 Life Technologies
LS12999442
100 Tests Each for $376.20
Specifications

Description

The dG9 antibody clone reacts with human perforin (pore-forming protein, pfp). Perforin is one of the cytolytic mediators present in the cytoplasmic granules of cytotoxic T lymphocytes (CTL) and natural killer cells (NK). Perforin is involved in the killing function by CTLs and NKs and has an important role in the immune response against tumors and virus infections.
Description & Specifications

Specifications

Perforin
Murine
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
dG9 (delta G9)
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer