Please login to your online account to display your discounted pricing

Perforin Mouse anti-Human, eFluor 450, Clone: dG9 (delta G9), eBioscience™

Mouse Monoclonal Antibody

$176.90 - $442.25

Specifications

Antigen Perforin
Clone dG9 (delta G9)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124241

 
life technologies
LS48999441
25 Tests Each for $176.90

501124242

 
life technologies
LS48999442
100 Tests Each for $442.25
Description & Specifications

Specifications

Antigen Perforin
Clone dG9 (delta G9)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The dG9 antibody clone reacts with human perforin (pore-forming protein, pfp). Perforin is one of the cytolytic mediators present in the cytoplasmic granules of cytotoxic T lymphocytes (CTL) and natural killer cells (NK). Perforin is involved in the killing function by CTLs and NKs and has an important role in the immune response against tumors and virus infections.