missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perforin Mouse anti-Human, Clone: dG9 (delta G9), eBioscience™

Mouse Monoclonal Antibody

$129.05 - $288.55

Specifications

Antigen Perforin
Clone dG9 (delta G9)
Host Species Murine
Species Reactivity Human
Applications Immunohistochemistry,Immunoprecipitation
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122900 Life Technologies
LS14999482
100μg Each for $288.55
5013977 Life Technologies
LS14999480
25μg Each for $129.05
Specifications

Description

The dG9 antibody clone reacts with human perforin (pore-forming protein, pfp). Perforin is one of the cytolytic mediators present in the cytoplasmic granules of cytotoxic T lymphocytes (CTL) and natural killer cells (NK). Perforin is involved in the killing function by CTLs and NKs and has an important role in the immune response against tumors and virus infections.
Description & Specifications

Specifications

Perforin
Murine
Immunohistochemistry,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
dG9 (delta G9)
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer