Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorotripentylamine, tech. 85%

$32.95 - $297.08

Chemical Identifiers

Chemical Name or Material Perfluorotripentylamine
CAS 338-84-1
Purity Grade Technical Grade
Molecular Formula [CF3(CF2)4]3N
Formula Weight 821.12
MDL Number MFCD00042367
Synonym Perfluorotriamylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1684806

  SDS
alfa aesar™
L1684806
5g Each for $32.95

AAL1684814

  SDS
alfa aesar™
L1684814
25g Each for $93.43

AAL1684822

  SDS
alfa aesar™
L1684822
100g Each for $297.08
Description & Specifications

Specifications

Formula Weight 821.12
Chemical Name or Material Perfluorotripentylamine
Molecular Formula [CF3(CF2)4]3N
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA TSCA
Purity Grade Technical Grade
Assay 85%
Boiling Point 210° to 220°C
Quantity 5g
MDL Number MFCD00042367
CAS 338-84-1
Synonym Perfluorotriamylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.