Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorotripentylamine, tech. 85%


Quantity 5g

Manufacturer: alfa aesar™  L1684806

 View more versions of this product

Catalog No. AAL1684806

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 821.12
Chemical Name or Material Perfluorotripentylamine
Molecular Formula [CF3(CF2)4]3N
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Synonym Perfluorotriamylamine
CAS 338-84-1
Purity Grade Technical Grade
Quantity 5g
Boiling Point 210° to 220°C
MDL Number MFCD00042367
Assay 85%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
TSCA TSCA
Health Hazard 1 Warning

Specifications

Chemical Name or Material Perfluorotripentylamine
CAS 338-84-1
Purity Grade Technical Grade
Molecular Formula [CF3(CF2)4]3N
Formula Weight 821.12
MDL Number MFCD00042367
Synonym Perfluorotriamylamine
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents