Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorotributylamine, Mass Spec Std

$37.68 - $292.31

Chemical Identifiers

Chemical Name or Material Perfluorotributylamine
CAS 311-89-7
Name Note Mass Spec Standard
Molecular Formula [CF3(CF2)3]3N
Formula Weight 671.1
MDL Number MFCD00000436
Beilstein 1813883
Synonym FC-43; Heptacosafluorotri-n-butylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1912603

  SDS
alfa aesar™
A1912603
1g Each for $37.68

AAA1912614

  SDS
alfa aesar™
A1912614
25g Each for $292.31
Description & Specifications

Specifications

Formula Weight 671.1
CAS 311-89-7
Boiling Point 177° to 180°C
Melting Point −52°C
Chemical Name or Material Perfluorotributylamine
Quantity 1g
TSCA TSCA
MDL Number MFCD00000436
Synonym FC-43; Heptacosafluorotri-n-butylamine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number YA1000000
Molecular Formula [CF3(CF2)3]3N
Beilstein 1813883
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Mass Spec Standard

Applications
Perfluorotributylamine is used in electronic applications viz. liquid burn-in, testing and vapor phase soldering processes. It acts as an indicator fluid to check the leakage and heat transfer fluid for thermal shock testing. It is an active component in fluosol, which is a substitute for artificial blood and fluorinert coolant liquids. Further, it is used as a solvent for computer disc drive lubrication. In addition to this, it is used as a calibration chemical in gas chromatography.

Solubility
Miscible with chloroform, cyclohexane, ethyl ether, ethyl acetate, heptane, carbon tetrachloride and petroleum ether. Immiscible with water, methanol, toluene, isopropyl alcohol, benzene and acetone.

Notes
Store in a cool place. Incompatible with powdered metals and alkali metals.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.