Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorotetradecane, 97%

$74.51 - $290.92

Chemical Identifiers

Chemical Name or Material Perfluorotetradecane
CAS 307-62-0
Molecular Formula C14F30
Linear Formula CF3(CF2)12CF3
Formula Weight 738.11
MDL Number MFCD00013571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1684603

  SDS
alfa aesar™
L1684603
1g Each for $74.51

AAL1684606

  SDS
alfa aesar™
L1684606
5g Each for $290.92
Description & Specifications

Specifications

Formula Weight 738.11
Chemical Name or Material Perfluorotetradecane
Molecular Formula C14F30
Linear Formula CF3(CF2)12CF3
MDL Number MFCD00013571
CAS 307-62-0
Assay 97%
Melting Point 103° to 105°C
Quantity 1g