missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfluorooctyl iodide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 269960010

 View more versions of this product

Catalog No. AC269960010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Perfluorooctyl iodide
>115°C
Glass bottle
2.04
Colorless to Yellow or Pink
1g
507-63-1
C8F17I
MFCD00001064
Solubility in water: insoluble
C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
545.96
545.96
98%
2.0400g/mL
Authentic
1.3286 to 1.3306
160°C to 161°C
25°C
99%
97.5% min. (GC)
CF3(CF2)7I
1-iodoheptadecafluorooctane, 1-iodoperfluorooctane, heptadecafluoro-1-iodooctane, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo, perfluoro-1-iodooctane, perfluoro-n-octyl iodide, perfluorooctyl iodide, perfluorooctyliodide
KWXGJTSJUKTDQU-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
10491
Liquid After Melting

Chemical Identifiers

507-63-1
545.96
KWXGJTSJUKTDQU-UHFFFAOYSA-N
10491
C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
C8F17I
MFCD00001064
1-iodoheptadecafluorooctane, 1-iodoperfluorooctane, heptadecafluoro-1-iodooctane, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo, perfluoro-1-iodooctane, perfluoro-n-octyl iodide, perfluorooctyl iodide, perfluorooctyliodide
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-079-5

RTECSNumber : RG9700050

TSCA : TSCA

SDS Documents