Learn More
Perfluorooctanesulfonamide, 95%
CAS: 754-91-6 | C8H2F17NO2S | 499.15 g/mol
$703.09 - $703.09
Chemical Identifiers
| CAS | 754-91-6 |
|---|---|
| Molecular Formula | C8H2F17NO2S |
| Molecular Weight (g/mol) | 499.15 |
| InChI Key | RRRXPPIDPYTNJG-UHFFFAOYSA-N |
| Synonym | perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide |
| PubChem CID | 69785 |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide |
| SMILES | C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC459640050
|
Thermo Scientific Chemicals
459640050 |
5 g |
Each for $703.09
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 754-91-6 | |
| 499.15 | |
| perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide | |
| 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide |
| C8H2F17NO2S | |
| RRRXPPIDPYTNJG-UHFFFAOYSA-N | |
| 69785 | |
| C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F |
Specifications
| 754-91-6 | |
| Authentic | |
| C8H2F17NO2S | |
| perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide | |
| C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F | |
| 499.15 | |
| 499.15 | |
| Perfluorooctanesulfonamide |
| 154.0°C to 155.0°C | |
| 94% min. (mix of isomers) (GC) | |
| 5 g | |
| RRRXPPIDPYTNJG-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide | |
| 69785 | |
| 95% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 212-046-
RUO – Research Use Only