Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorooctadecanoic acid, 97%

Chemical Identifiers

Chemical Name or Material Perfluorooctadecanoic acid
CAS 16517-11-6
Molecular Formula C18HF35O2
Linear Formula CF3(CF2)16CO2H
Formula Weight 914.15
MDL Number MFCD00198021
UN Number UN3261
Synonym Perfluorostearic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1683706

 
alfa aesar™
L1683706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1683714

 
alfa aesar™
L1683714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 914.15
Chemical Name or Material Perfluorooctadecanoic acid
Assay 97%
CAS 16517-11-6
TSCA TSCA
Boiling Point 235°C (100mmHg)
Melting Point 162° to 172°C
Molecular Formula C18HF35O2
Quantity 5g
Linear Formula CF3(CF2)16CO2H
MDL Number MFCD00198021
UN Number UN3261
Synonym Perfluorostearic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
Perfluorooctadecanoic acid is used to form thin films by self-assembly on the surface of Chromium-Nickel stainless steel.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.