Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorononanoic acid, 97%

Chemical Identifiers

Chemical Name or Material Perfluorononanoic acid
CAS 375-95-1
Molecular Formula C9HF17O2
Linear Formula CF3(CF2)7CO2H
Formula Weight 464.07
MDL Number MFCD00039605
UN Number UN3261
Beilstein 1897287
Synonym n-Heptadecafluorononanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1398006

  SDS
alfa aesar™
A1398006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1398014

  SDS
alfa aesar™
A1398014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1398022

  SDS
alfa aesar™
A1398022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 464.07
Chemical Name or Material Perfluorononanoic acid
TSCA TSCA
Melting Point 63° to 68°C
Quantity 5g
Linear Formula CF3(CF2)7CO2H
MDL Number MFCD00039605
Health Hazard 1 Danger
Assay 97%
Boiling Point 218°C
Molecular Formula C9HF17O2
CAS 375-95-1
UN Number UN3261
Beilstein 1897287
Synonym n-Heptadecafluorononanoic acid
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.