Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorokerosene, low boiling

Chemical Identifiers

Chemical Name or Material Perfluorokerosene
Name Note low boiling
MDL Number MFCD00132835
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL10538MD

  SDS
alfa aesar™
L10538MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Perfluorokerosene
Name Note low boiling
MDL Number MFCD00132835
Boiling Point ≈40° to 100°C
Quantity 250mg