Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorokerosene-L

$455.62 - $455.62

Chemical Identifiers

Chemical Name or Material Perfluorokerosene-L
MDL Number MFCD00132835
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1659703

 
alfa aesar™
L1659703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1659706

 
alfa aesar™
L1659706
5g Each for $455.62
Description & Specifications

Specifications

Chemical Name or Material Perfluorokerosene-L
MDL Number MFCD00132835
Boiling Point 70° to 200°C
Quantity 1g

Applications
Perfluorokerosene-L is used as a standard in mass spectrometry. It is used as a solvent in the preparation of 1,3-oxazin-2-ones and oxazolidin-2-ones.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.