Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorokerosene-941

$272.45 - $272.45

Chemical Identifiers

Chemical Name or Material Perfluorokerosene-941
MDL Number MFCD00132835
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1659503

 
alfa aesar™
L1659503
1g Each for $272.45
Description & Specifications

Specifications

Chemical Name or Material Perfluorokerosene-941
MDL Number MFCD00132835
Boiling Point 110° to 180°C (0.5mmHg)
Quantity 1g

Applications
Perfluorokerosene is used as internal reference for negative ion mass spectrometry, mulling agent during the analysis of urinary calculi by diffuse reflectance infrared Fourier transform spectroscopy.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.