missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorohexanes, 98+%

Manufacturer:  Alfa Aesar™ 1581236

 View more versions of this product

Catalog No. AA1581236



Specifications

Specifications

Perfluorohexanes
1.2510
Colorless
500g
C6F14
MFCD00000437
perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
338.044
CHEBI:39427
Liquid
1.699g/mL
58°C to 60°C
-4°C
355-42-0
CF3(CF2)4CF3
1802113
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
9639
338.04
≥98%

Chemical Identifiers

355-42-0
338.044
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
9639
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
C6F14
MFCD00000437
perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
CHEBI:39427
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MO4310000

TSCA : TSCA

SDS Documents