missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoroeicosane, 97%

$418.69 - $418.69

Chemical Identifiers

CAS 37589-57-4
Molecular Formula CF3(CF2)18CF3
Formula Weight 1038.16
MDL Number MFCD00000438
Chemical Name or Material Perfluoroeicosane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1682806 SDS Alfa Aesar™
L1682806
5g Each for $418.69
Specifications Description & Specifications

Chemical Identifiers

37589-57-4
1038.16
Perfluoroeicosane
CF3(CF2)18CF3
MFCD00000438

Specifications

37589-57-4
162° to 165°C
5g
MFCD00000438
97%
320°C
CF3(CF2)18CF3
1038.16
Perfluoroeicosane