missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorododecane, 97%

$91.14 - $309.96

Chemical Identifiers

CAS 307-59-5
Molecular Formula C12F26
Molecular Weight (g/mol) 638.09
MDL Number MFCD00042084
InChI Key WNZGTRLARPEMIG-UHFFFAOYSA-N
Synonym perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2
PubChem CID 78973
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecane
SMILES C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1682703 View Documents Alfa Aesar™
L1682703
1g Each for $91.14
 
AAL1682706 View Documents Alfa Aesar™
L1682706
5g Each for $309.96
 
Specifications

Chemical Identifiers

307-59-5
638.09
WNZGTRLARPEMIG-UHFFFAOYSA-N
78973
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C12F26
MFCD00042084
perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecane

Specifications

178°C
1g
C12F26
MFCD00042084
perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2
C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
638.09
638.09
Perfluorododecane
72°C to 76°C
307-59-5
CF3(CF2)10CF3
1811139
WNZGTRLARPEMIG-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecane
78973
97%