Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluorodimethylcyclobutane, mixture of 1,2 and 1,3 isomers, 97%

$50.53 - $50.53

Chemical Identifiers

Chemical Name or Material Perfluorodimethylcyclobutane
CAS 28677-00-1
Name Note Mixture of 1,2 and 1,3 isomers
Molecular Formula C6F12
Linear Formula C4F6(CF3)2
Formula Weight 300.05g/mol
MDL Number MFCD00013737
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4303609

 
alfa aesar™
43036-09
10g Each for $50.53
Description & Specifications

Specifications

Density 1.620g/mL
Refractive Index 1.2540
Boiling Point 45°C
Quantity 10g
Chemical Name or Material Perfluorodimethylcyclobutane
Name Note Mixture of 1,2 and 1,3 isomers
CAS 28677-00-1
Physical Form Liquid
Assay 97%
Molecular Formula C6F12
Linear Formula C4F6(CF3)2
Formula Weight 300.05g/mol
MDL Number MFCD00013737