Learn More
Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
$108.48 - $386.96
Chemical Identifiers
CAS | 335-76-2 |
---|---|
Molecular Formula | C10HF19O2 |
Molecular Weight (g/mol) | 514.09 |
MDL Number | MFCD00004175 |
InChI Key | PCIUEQPBYFRTEM-UHFFFAOYSA-N |
Synonym | perfluorodecanoic acid, nonadecafluorodecanoic acid, ndfda, pfda, perfluoro-n-decanoic acid, nonadecafluoro-n-decanoic acid, decanoic acid, nonadecafluoro, ccris 4417, decanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro, perfluorocapric acid |
PubChem CID | 9555 |
ChEBI | CHEBI:35546 |
IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid |
SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2229206
|
Thermo Scientific Chemicals
B2229206 |
5 g |
Each for $108.48
|
|
|||||
AAB2229214
|
Thermo Scientific Chemicals
B2229214 |
25 g |
Each for $386.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
335-76-2 | |
514.09 | |
PCIUEQPBYFRTEM-UHFFFAOYSA-N | |
9555 | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid |
C10HF19O2 | |
MFCD00004175 | |
perfluorodecanoic acid, nonadecafluorodecanoic acid, ndfda, pfda, perfluoro-n-decanoic acid, nonadecafluoro-n-decanoic acid, decanoic acid, nonadecafluoro, ccris 4417, decanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro, perfluorocapric acid | |
CHEBI:35546 | |
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
Specifications
335-76-2 | |
218°C | |
CF3(CF2)8CO2H | |
5 g | |
1810811 | |
PCIUEQPBYFRTEM-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid | |
9555 | |
514.09 | |
Perfluorodecanoic acid |
77°C to 82°C | |
C10HF19O2 | |
MFCD00004175 | |
UN2923 | |
perfluorodecanoic acid, nonadecafluorodecanoic acid, ndfda, pfda, perfluoro-n-decanoic acid, nonadecafluoro-n-decanoic acid, decanoic acid, nonadecafluoro, ccris 4417, decanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro, perfluorocapric acid | |
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | |
514.09 | |
CHEBI:35546 | |
97% |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H351-H360Df-H362
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 206-400-3
RTECSNumber : HD9900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only