missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perfluorodecalin, 90%, mixture of cis and trans, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130041000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Perfluorodecalin | |
-10.0°C | |
1.9000g/mL | |
59°C | |
88% min. (GC) | |
C10F18 | |
MFCD00010626 | |
1.9 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene | |
9386 | |
462.08 | |
Liquid |
306-94-5 | |
Colorless | |
142.0°C | |
Authentic | |
Glass bottle | |
1.3135 to 1.3155 | |
100 g | |
15,4215 | |
Solubility in water: insoluble | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F | |
462.08 | |
CHEBI:38848 | |
90% |
Chemical Identifiers
306-94-5 | |
462.08 | |
UWEYRJFJVCLAGH-UHFFFAOYSA-N | |
9386 | |
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene |
C10F18 | |
MFCD00010626 | |
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin | |
CHEBI:38848 | |
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Warning
EINECSNumber : 206-192-4
RTECSNumber : QJ3175000
TSCA : TSCA
RUO – Research Use Only