missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 100ppm 4-methoxyphenol

Manufacturer:  Alfa Aesar™ 4479314

 View more versions of this product

Catalog No. AA4479314



Specifications

Specifications

(Perfluorocyclohexyl)methyl acrylate
Colorless
Stabilized with 100ppm 4-methoxyphenol
C10H5F11O2
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F
366.13
366.13
96%
Air Sensitive
25g
40677-94-9
MFCD00080614
OZGWOALFBHODRB-UHFFFAOYSA-N
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate
2776049
Liquid

Chemical Identifiers

40677-94-9
366.13
OZGWOALFBHODRB-UHFFFAOYSA-N
2776049
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F
C10H5F11O2
MFCD00080614
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS Documents