missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04479306
Description
(Perfluorocyclohexyl)methyl acrylate is used in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(Perfluorocyclohexyl)methyl acrylate | |
40677-94-9 | |
C10H5F11O2 | |
5 g | |
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester | |
OZGWOALFBHODRB-UHFFFAOYSA-N | |
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate | |
2776049 | |
96% |
Stabilized with 100ppm 4-methoxyphenol | |
Colorless | |
MFCD00080614 | |
Air sensitive | |
Sparingly soluble in water. | |
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F | |
366.13 | |
366.13 | |
Liquid |
Chemical Identifiers
40677-94-9 | |
366.13 | |
OZGWOALFBHODRB-UHFFFAOYSA-N | |
2776049 | |
C=CC(=O)OCC1(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(F)F)F |
C10H5F11O2 | |
MFCD00080614 | |
perfluorocyclohexylmethyl acrylate, perfluorocyclohexyl methyl acrylate, undecafluorocyclohexyl methyl prop-2-enoate, 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl prop-2-enoate, acrylic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexylmethyl ester, 1,2,2,3,3,4,4,5,5,6,6-undecakis fluoranyl cyclohexyl methyl prop-2-enoate, 2-propenoic acid 1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl methyl ester | |
(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)methyl prop-2-enoate |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only