missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perfluorobutyl iodide, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC269951000
Specifications
Perfluorobutyl iodide | |
>115°C | |
Glass bottle | |
2.01 | |
Purple | |
100g | |
98.5% min. (GC) | |
CF3(CF2)3I | |
1-iodononafluorobutane, 1-iodoperfluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo, ccris 9008, n-nonafluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluoro-n-butyl iodide, perfluorobutyl iodide, perfluorobutyliodide | |
PGRFXXCKHGIFSV-UHFFFAOYSA-N | |
1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane | |
67917 | |
Liquid |
2.0100g/mL | |
Authentic | |
1.3275 to 1.3295 | |
64°C to 66°C (973.0 mbar) | |
-88°C | |
423-39-2 | |
C4F9I | |
MFCD00001062 | |
Solubility in water: immiscible | |
C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F | |
345.92 | |
345.92 | |
99% |
Chemical Identifiers
423-39-2 | |
345.92 | |
PGRFXXCKHGIFSV-UHFFFAOYSA-N | |
67917 | |
C(C(C(F)(F)I)(F)F)(C(F)(F)F)(F)F |
C4F9I | |
MFCD00001062 | |
1-iodononafluorobutane, 1-iodoperfluorobutane, butane, 1,1,1,2,2,3,3,4,4-nonafluoro-4-iodo, ccris 9008, n-nonafluorobutyl iodide, nonafluoro-1-iodobutane, nonafluorobutyl iodide, perfluoro-n-butyl iodide, perfluorobutyl iodide, perfluorobutyliodide | |
1,1,1,2,2,3,3,4,4-nonafluoro-4-iodobutane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
EINECSNumber : 207-025-8