missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoroazelaic acid, tech. 90%

$82.96 - $279.12

Chemical Identifiers

CAS 23453-64-7
Molecular Formula C9H2F14O4
Molecular Weight (g/mol) 440.089
MDL Number MFCD00153274
InChI Key WRYSWXOLGMKBKW-UHFFFAOYSA-N
Synonym perfluoroazelaic acid, tetradecafluoroazelaic acid, perfluorononanedioic acid, acmc-20ajy5, tetradecafluorononanedioic acid, perfluorononane-1,9-dioic acid, perfluorononane-1,9-dioic acid, tech, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonanedioic acid, nonanedioic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro
PubChem CID 2776030
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononanedioic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1682006 View Documents Alfa Aesar™
L1682006
5g Each for $82.96
 
AAL1682014 View Documents Alfa Aesar™
L1682014
25g Each for $279.12
 
Specifications

Chemical Identifiers

23453-64-7
440.089
WRYSWXOLGMKBKW-UHFFFAOYSA-N
2776030
C(=O)(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
C9H2F14O4
MFCD00153274
perfluoroazelaic acid, tetradecafluoroazelaic acid, perfluorononanedioic acid, acmc-20ajy5, tetradecafluorononanedioic acid, perfluorononane-1,9-dioic acid, perfluorononane-1,9-dioic acid, tech, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonanedioic acid, nonanedioic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononanedioic acid

Specifications

215°C (60mmHg)
23453-64-7
MFCD00153274
3020403
WRYSWXOLGMKBKW-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononanedioic acid
2776030
90%
Perfluoroazelaic acid
5g
C9H2F14O4
UN3261
perfluoroazelaic acid, tetradecafluoroazelaic acid, perfluorononanedioic acid, acmc-20ajy5, tetradecafluorononanedioic acid, perfluorononane-1,9-dioic acid, perfluorononane-1,9-dioic acid, tech, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonanedioic acid, nonanedioic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro
C(=O)(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
440.089
440.09
Technical
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.