missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfluoro(decahydronaphthalene), cis + trans, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1828836

 View more versions of this product

Catalog No. AAA1828836



Description

Description

Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(decahydronaphthalene) is used to dissolve gases. Due to its gas carrying capacity, it is utilized to increase oxygen delivery during cell culture. It acts as an ingredient in fluosol which is an artificial blood product. It is also used to dissolve Teflon.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Perfluoro(decahydronaphthalene)
306-94-5
1.94
C10F18
MFCD00010626
2067113
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
UWEYRJFJVCLAGH-UHFFFAOYSA-N
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
9386
462.1
cis + trans
-10°C to -7°C
141°C to 142°C
1.3145
500 g
14,4183
Insoluble in water.
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F
462.081
CHEBI:38848
95%

Chemical Identifiers

306-94-5
462.081
UWEYRJFJVCLAGH-UHFFFAOYSA-N
9386
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
C10F18
MFCD00010626
perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
CHEBI:38848
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 206-192-4

RTECSNumber : QJ3175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only