Please login to your online account to display your discounted pricing

Perfluoro-compound FC-770™, ACROS Organics™


Quantity 50mL

Manufacturer: thermo fisher scientific  AC394890500

 View more versions of this product

Catalog No. AC394890500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 50mL
Density 1.793g/cm3
Chemical Name or Material Perfluoro-compound FC-770™
MDL Number MFCD09753882
CAS 1093615-61-2
Synonym Fluorinert™ FC-770

Specifications

Chemical Name or Material Perfluoro-compound FC-770™
CAS 1093615-61-2
MDL Number MFCD09753882
Synonym Fluorinert™ FC-770