missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perfluoro-compound FC-770(TM), Thermo Scientific Chemicals
$103.96 - $163.98
Chemical Identifiers
CAS | 1093615-61-2 |
---|---|
Molecular Weight (g/mol) | 399 |
MDL Number | MFCD09753882 |
Synonym | Fluorinert™ FC-770 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC394890250
|
Thermo Scientific Chemicals
394890250 |
25 mL |
Each for $103.96
|
|
|||||
AC394890500
|
Thermo Scientific Chemicals
394890500 |
50 mL |
Each for $163.98
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1093615-61-2 | |
399 | |
MFCD09753882 | |
Fluorinert™ FC-770 |
Specifications
1093615-61-2 | |
95.0°C | |
MFCD09753882 | |
1.8 | |
Solubility in water: insoluble | |
0.8 mm2/s (25°C) | |
Perfluoro-compound FC-770* |
1.8000g/mL | |
Glass bottle | |
25 mL | |
Fluorinert™ FC-770 | |
399 | |
399 |
RUO – Research Use Only