missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfluoro-compound FC-72(TM), ACROS Organics™

$191.65 - $399.49

Chemical Identifiers

CAS 355-42-0
Molecular Formula C6F14
Molecular Weight (g/mol) 340
MDL Number MFCD00000437
InChI Key ZJIJAJXFLBMLCK-UHFFFAOYSA-N
Synonym fluorinert fc72, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, perflexane, perflexane usan:inn, perfluoro-n-hexane, perfluorohexane, perfluorohexanes, tetradecafluorohexane
PubChem CID 9639
ChEBI CHEBI:39427
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC123791000
SDS View Documents
Thermo Fisher Scientific
123791000
100g Glass bottle
Each for $191.65
 
AC123792500
SDS View Documents
Fisher Scientific Acros
123792500
250g Glass bottle
Each for $399.49
 
Description

Description

Specifications

Chemical Identifiers

355-42-0
340
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
9639
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
C6F14
MFCD00000437
fluorinert fc72, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, perflexane, perflexane usan:inn, perfluoro-n-hexane, perfluorohexane, perfluorohexanes, tetradecafluorohexane
CHEBI:39427
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

1.6800g/mL
Glass bottle
1.68
51.0°C to 59.0°C
-4.0°C
355-42-0
MFCD00000437
fluorinert fc72, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, perflexane, perflexane usan:inn, perfluoro-n-hexane, perfluorohexane, perfluorohexanes, tetradecafluorohexane
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
9639
340
Perfluoro-compound FC-72&asterisk;
Authentic
1.251
0.42 mm2/s (25°C)
Colorless
100g
C6F14
15, 7271
Solubility in water: insoluble
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
340
CHEBI:39427
Liquid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause long lasting harmful effects to aquatic life.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-585-0

RTECSNumber : MO4310000

TSCA : TSCA

SDS
Product Certifications