Please login to your online account to display your discounted pricing

Perfluoro-Compound FC-43, ACROS Organics™

$103.07 - $103.07

Chemical Identifiers

Chemical Name or Material Perfluoro-compound FC-43™
CAS 311-89-7
Molecular Formula C12F27N
Formula Weight 670
MDL Number MFCD00000436
Synonym Fluorinert™ FC-43
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC123770100

  SDS
thermo fisher scientific
123770100
Glass bottle 10g Each for $103.07

AC123771000

  SDS
fisher scientific acros
123771000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC123772500

  SDS
fisher scientific acros
123772500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Boiling Point 173° to 181°C
Formula Weight 670
Quantity 10g
Color Clear colorless
Density 1.88g/mL
Viscosity 2.3 to 3.3 cSt (25°C)
EINECS Number 206-223-1
RTECS Number YA1000000
TSCA TSCA
Distillation Range distillation range (between 165 and 185°C):> 97%
Synonym Fluorinert™ FC-43
Infrared Spectrum Authentic
Packaging Glass bottle
Pour Point pour point: < - 40°C
Dielectric Strength > 35 kV
Distillation Residue residue: < 10 μg/mL
MDL Number MFCD00000436
CAS 311-89-7
Chemical Name or Material Perfluoro-compound FC-43™
Molecular Formula C12F27N