Please login to your online account to display your discounted pricing

Perfluoro-Compound FC-43, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  123772500

 View more versions of this product

Catalog No. AC123772500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Boiling Point 173° to 181°C
Formula Weight 670
Physical Form Liquid
Quantity 250g
Color Clear colorless
Density 1.88g/mL
Viscosity 2.3 to 3.3 cSt (25°C)
RTECS Number YA1000000
EINECS Number 206-223-1
TSCA TSCA
Chemical Name or Material Perfluoro-compound FC-43™
Pour Point pour point: < - 40°C
Dielectric Strength > 35 kV
Molecular Formula C12F27N
MDL Number MFCD00000436
CAS 311-89-7
Distillation Range distillation range (between 165 and 185°C):> 97%
Synonym Fluorinert™ FC-43
Distillation Residue residue: < 10 μg/mL
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material Perfluoro-compound FC-43™
CAS 311-89-7
Molecular Formula C12F27N
Formula Weight 670
MDL Number MFCD00000436
Synonym Fluorinert™ FC-43