Please login to your online account to display your discounted pricing

Perfluoro-compound FC-40(TM), ACROS Organics™

$149.51 - $306.39

Chemical Identifiers

Chemical Name or Material Perfluoro-compound FC-40™
CAS 51142-49-5
Formula Weight 650
MDL Number MFCD01632188
Synonym Fluorinert™ FC-40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC123761000

  SDS
fisher scientific acros
123761000
Glass bottle 100g Each for $149.51

AC123762500

  SDS
fisher scientific acros
123762500
Glass bottle 250g Each for $306.39

AC123760100

  SDS
thermo fisher scientific
AC123760100
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 158° to 173°C
Physical Form Liquid
Formula Weight 650
Quantity 100g
Color Clear colorless
Density 1.8700g/mL
Viscosity 1.6-2.8cSt (25°C)
Distillation Range distillation range (between 139 and 189°C):> 95%
Distillation Residue residue: < 10 μg/mL
Synonym Fluorinert™ FC-40
Packaging Glass bottle
Pour Point pour point < -50°C
Infrared Spectrum Authentic
Dielectric Strength > 35 kV
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause long lasting harmful effects to aquatic life.
CAS 51142-49-5
Chemical Name or Material Perfluoro-compound FC-40™
MDL Number MFCD01632188
Safety and Handling

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause long lasting harmful effects to aquatic life.