Please login to your online account to display your discounted pricing

Perfluoro-compound FC-40(TM), ACROS Organics™


Quantity 10g

Manufacturer: thermo fisher scientific  AC123760100

 View more versions of this product

Catalog No. AC123760100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Boiling Point 158° to 173°C
Formula Weight 650
Quantity 10g
Color Clear colorless
Density 1.8700g/mL
Viscosity 1.6-2.8cSt (25°C)
Synonym Fluorinert™ FC-40
Pour Point pour point < -50°C
Dielectric Strength > 35 kV
Distillation Range distillation range (between 139 and 189°C):> 95%
Health Hazard 2 GHS H Statement:
May cause long lasting harmful effects to aquatic life.
CAS 51142-49-5
Distillation Residue residue: < 10 μg/mL
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Perfluoro-compound FC-40™
MDL Number MFCD01632188

Specifications

Chemical Name or Material Perfluoro-compound FC-40™
CAS 51142-49-5
Formula Weight 650
MDL Number MFCD01632188
Synonym Fluorinert™ FC-40
Accessories

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause long lasting harmful effects to aquatic life.

SDS & Safety Documents