missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Perfluoro(allylbenzene), 97%, Thermo Scientific Chemicals
$90.52 - $332.37
Chemical Identifiers
CAS | 67899-41-6 |
---|---|
Molecular Formula | C9F10 |
Molecular Weight (g/mol) | 298.083 |
MDL Number | MFCD00153225 |
InChI Key | WRHBYJDZKRNITP-UHFFFAOYSA-N |
Synonym | perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene |
PubChem CID | 2775989 |
IUPAC Name | 1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene |
SMILES | C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2146906
|
Thermo Scientific Chemicals
B2146906 |
5 g |
Each for $90.52
|
|
|||||
AAB2146914
|
Thermo Scientific Chemicals
B2146914 |
25 g |
Each for $332.37
|
|
|||||
Description
Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPerfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, ammonia.
Chemical Identifiers
67899-41-6 | |
298.083 | |
WRHBYJDZKRNITP-UHFFFAOYSA-N | |
2775989 | |
C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F |
C9F10 | |
MFCD00153225 | |
perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene | |
1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene |
Specifications
67899-41-6 | |
C9F10 | |
5 g | |
Insoluble in water. | |
C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F | |
298.083 | |
298.08 | |
Perfluoro(allylbenzene) |
1.708 | |
MFCD00153225 | |
perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene | |
WRHBYJDZKRNITP-UHFFFAOYSA-N | |
1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene | |
2775989 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only