missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perfluoro(allylbenzene), 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2146906

 View more versions of this product

Catalog No. AAB2146906



Description

Description

Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro(allylbenzene) is used as a pharmaceutical intermediate and as a chemical in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, ammonia.
Specifications

Specifications

Perfluoro(allylbenzene)
1.708
MFCD00153225
perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene
WRHBYJDZKRNITP-UHFFFAOYSA-N
1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene
2775989
97%
67899-41-6
C9F10
5 g
Insoluble in water.
C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F
298.083
298.08

Chemical Identifiers

67899-41-6
298.083
WRHBYJDZKRNITP-UHFFFAOYSA-N
2775989
C1(=C(C(=C(C(=C1F)F)F)F)F)C(C(=C(F)F)F)(F)F
C9F10
MFCD00153225
perfluoro allylbenzene, perfluoroallylbenzene, 3-pentafluorophenyl pentafluoro-1-propene, 3-pentafluorophenyl pentafluoroprop-1-ene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-enyl benzene, 1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoroprop-2-en-1-yl benzene, 1,2,3,4,5-pentafluoro-6-pentafluoroprop-2-en-1-yl benzene, benzene,1,2,3,4,5-pentafluoro-6-1,1,2,3,3-pentafluoro-2-propen-1-yl, acmc-1b8oa, 1-pentafluoroallyl pentafluorobenzene
1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-enyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only