missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoro(4-methylmorpholine), 98%

$59.53 - $59.53

Chemical Identifiers

CAS 382-28-5
Molecular Formula C5F11NO
Formula Weight 299.04
MDL Number MFCD00042442
Synonym 2,2,3,3,5,5,6,6-Octafluoro-4-(trifluoromethyl)morpholine
Chemical Name or Material Perfluoro(4-methylmorpholine)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2135806 SDS Alfa Aesar™
B2135806
5g Each for $59.53
AAB2135814 SDS Alfa Aesar™
B2135814
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

382-28-5
299.04
2,2,3,3,5,5,6,6-Octafluoro-4-(trifluoromethyl)morpholine
C5F11NO
MFCD00042442
Perfluoro(4-methylmorpholine)

Specifications

382-28-5
5g
MFCD00042442
Perfluoro(4-methylmorpholine)
C5F11NO
299.04
2,2,3,3,5,5,6,6-Octafluoro-4-(trifluoromethyl)morpholine
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA