Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoro(2-n-butyltetrahydrofuran), tech., cont. perfluoro(2-n-propyltetrahydropyran)

$149.18 - $616.27

Chemical Identifiers

Chemical Name or Material Perfluoro(2-n-butyltetrahydrofuran)
CAS 335-36-4
Purity Grade Technical Grade
Molecular Formula C8F16O
Formula Weight 416.06
MDL Number MFCD00014513
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2009818

  SDS
alfa aesar™
B2009818
50g Each for $149.18

AAB2009830

  SDS
alfa aesar™
B2009830
250g Each for $616.27
Description & Specifications

Specifications

Formula Weight 416.06
Chemical Name or Material Perfluoro(2-n-butyltetrahydrofuran)
TSCA TSCA
Purity Grade Technical Grade
Boiling Point 99° to 107°C
Assay Percent Notes cont. perfluoro(2-n-propyltetrahydropyran)
Molecular Formula C8F16O
Quantity 50g
MDL Number MFCD00014513
CAS 335-36-4

Applications
It is a reaction medium with substantial capacity to solubilize molecular oxygen as in the high yield epoxidation of alkenes in the presence of trimethylacetaldehyde.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.