Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride, 97%

Chemical Identifiers

Chemical Name or Material Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride
CAS 27639-98-1
Molecular Formula C12F24O4
Formula Weight 664.09
MDL Number MFCD00084743
UN Number UN3265
Synonym HFPO tetramer acid fluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1661406

  SDS
alfa aesar™
L1661406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1661414

  SDS
alfa aesar™
L1661414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 664.09
Chemical Name or Material Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride
CAS 27639-98-1
MDL Number MFCD00084743
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12F24O4
Assay 97%
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Health Hazard 1 Danger
Synonym HFPO tetramer acid fluoride
Quantity 5g
UN Number UN3265
Boiling Point 158° to 161°C
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.