Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoro-1-iodohexane, 98+%, stab. with copper

$55.27 - $840.45

Chemical Identifiers

Chemical Name or Material Perfluoro-1-iodohexane
CAS 355-43-1
Name Note stabilized with copper
Molecular Formula CF3(CF2)5I
Formula Weight 445.95
MDL Number MFCD00001063
Beilstein 1802118
Synonym 1-Iodoperfluorohexane; Perfluoro-n-hexyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2095809

 
alfa aesar™
B2095809
10g Each for $55.27

AAB2095818

 
alfa aesar™
B2095818
50g Each for $212.42

AAB2095830

 
alfa aesar™
B2095830
250g Each for $840.45
Description & Specifications

Specifications

Formula Weight 445.95
Chemical Name or Material Perfluoro-1-iodohexane
Boiling Point 117°C
TSCA TSCA
Name Note stabilized with copper
Melting Point −45°C
Assay ≥98%
Molecular Formula CF3(CF2)5I
MDL Number MFCD00001063
CAS 355-43-1
Quantity 10g
Beilstein 1802118
Synonym 1-Iodoperfluorohexane; Perfluoro-n-hexyl iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.