missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perfluoro(1,3-dimethylcyclohexane), tech. 80%, remainder other fluorocarbons

$89.61 - $296.19

Chemical Identifiers

CAS 335-27-3
Molecular Formula C8F16
Formula Weight 400.06
MDL Number MFCD00001469
Beilstein 2064147
Chemical Name or Material Perfluoro(1,3-dimethylcyclohexane)
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0572614 SDS Alfa Aesar™
L0572614
25g Each for $89.61
AAL0572622 SDS Alfa Aesar™
L0572622
100g Each for $296.19
Specifications Description & Specifications

Chemical Identifiers

335-27-3
400.06
2064147
Technical Grade
C8F16
MFCD00001469
Perfluoro(1,3-dimethylcyclohexane)

Specifications

335-27-3
−70°C
25g
MFCD00001469
Perfluoro(1,3-dimethylcyclohexane)
Technical Grade
101° to 102°C
C8F16
400.06
2064147
80%, remainder other fluorocarbons

Safety and Handling

RTECSNumber : GV4990000