Please login to your online account to display your discounted pricing

Honeywell Perfect Fit Kevlar Sleeves

$383.47 - $383.47

Specifications

Color Black
Material Kevlar™
For Use With (Application) Metal sheet handling, recycling, automotive assembly, blade handling, knife sharpening, Glass handling
Thickness 2-ply
View More Specs
Products
Catalog Number Mfr. No. Includes Length (English) Price Quantity    

19-050-008

 
honeywell
KVS214TH3BT
Thumbhole; 3 bar-tack for fingers 14 in. Case of 100 Each for $383.47
Description & Specifications

Specifications

Color Black
Material Kevlar™
For Use With (Application) Metal sheet handling, recycling, automotive assembly, blade handling, knife sharpening, Glass handling
Thickness 2-ply