Learn More
Perchloromethyl mercaptan, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 130030050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Perchloromethyl mercaptan | |
594-42-3 | |
146.0°C to 148.0°C | |
97% | |
CCl4S | |
CCl3SCl | |
5g | |
1.7 | |
RYFZYYUIAZYQLC-UHFFFAOYSA-N | |
trichloromethyl thiohypochlorite | |
11666 | |
97% |
97% | |
1.7000g/mL | |
Authentic | |
Glass bottle | |
1.5370 to 1.5410 | |
MFCD00037054 | |
03,135 | |
perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride | |
ClSC(Cl)(Cl)Cl | |
185.87 | |
185.88 |
Chemical Identifiers
594-42-3 | |
185.87 | |
RYFZYYUIAZYQLC-UHFFFAOYSA-N | |
11666 | |
ClSC(Cl)(Cl)Cl |
CCl4S | |
MFCD00037054 | |
perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride | |
trichloromethyl thiohypochlorite |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 209-840-4
RTECSNumber : PB0370000
TSCA : TSCA
RUO â Research Use Only