Please login to your online account to display your discounted pricing

Alfa Aesar™ Perchloric acid, Environmental Grade Plus, 65-71%

Chemical Identifiers

Chemical Name or Material Perchloric acid
CAS 7601-90-3
Purity Grade Environmental Grade Plus
Formula Weight 100.46
MDL Number MFCD00011325
UN Number UN1873
Merck Index 14,7153
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38750AP

 
alfa aesar™
38750AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Double Sub-Boiling Quartz Distillation
Formula Weight 100.46
Chemical Name or Material Perchloric acid
TSCA TSCA
Flash Point 113° (235°F)
Melting Point −12°C
Boiling Point 203°C
Purity Grade Environmental Grade Plus
Assay 65 to 71%
MDL Number MFCD00011325
CAS 7601-90-3
UN Number UN1873
Merck Index 14,7153
Density 1.67g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID
Odor Odorless
RTECS Number SC7500000
Color Colorless
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.