missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Perchloric acid, Environmental Grade, 67-71%

$299.18 - $842.00

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.454
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES OCl(=O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38749AP View Documents Alfa Aesar™
38749AP
500mL Each for $299.18
 
AA38749K4 View Documents Alfa Aesar™
38749K4
2.5L Each for $842.00
 
Specifications

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
hclo4
CHEBI:29221
OCl(=O)(=O)=O

Specifications

Colorless
7601-90-3
MFCD00011325
14,7153
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
100.454
CHEBI:29221
Liquid, Single Sub-Boiling Quartz Distillation
Environmental
113° (235°F)
203°C
500mL
ClHO4
UN1873
hclo4
OCl(=O)(=O)=O
1.67g/mL
24247
100.46
65 to 71%
Odorless
-12°C
Perchloric acid
Safety and Handling

Safety and Handling

GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID

RTECSNumber : SC7500000

TSCA : TSCA