missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric Acid 70% AR (ACS), Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.454
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES OCl(=O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK2766500 View Documents Avantor (Macron)
276614
500mL Glass N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK8828500 View Documents Avantor (Macron)
882814
500mL Glass N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK3999212 View Documents Avantor (Macron)
399946
2.5L SAFEMOR N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7601-90-3
100.454
hclo4
CHEBI:29221
OCl(=O)(=O)=O
ClHO4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid

Specifications

500mL
60%
hclo4
OCl(=O)(=O)=O
100.454
CHEBI:29221
Liquid
Glass
7601-90-3
ClHO4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
ACS
Perchloric Acid