missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric acid, ACS, 70%, redistilled, Alfa Aesar™

Manufacturer:  Alfa Aesar 87963K4

 View more versions of this product

Catalog No. AA87963K4



Specifications

Specifications

Perchloric acid
2.5L
7601-90-3
70%
ClHO4
UN1873
VLTRZXGMWDSKGL-UHFFFAOYSA-N
OCl(=O)(=O)=O
100.454
CHEBI:29221
Liquid
113°C (235°F)
203°C
Colorless
Redistilled
ACS Grade
70%
MFCD00011325
14,7153
1.67g/mL
perchloric acid
24247
100.46
Odorless
-112°C

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
CHEBI:29221
OCl(=O)(=O)=O
ClHO4
MFCD00011325
24247
perchloric acid
Safety and Handling

Safety and Handling

GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID

RTECSNumber : SC7500000

TSCA : TSCA

SDS Documents