missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric acid, ACS, 48-50%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm

Manufacturer:  Thermo Fisher Scientific 44464K4

 View more versions of this product

Catalog No. AA44464K4



Specifications

Specifications

Perchloric acid
7601-90-3
ClHO4
UN1802
Miscible with water.
OCl(=O)(=O)=O
1.41
24247
100.46
ACS
°C to 112°C
2.5L
48-50%
MFCD00011325
hclo4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
100.454
CHEBI:29221
Liquid
29mm Hg at 20°C

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
hclo4
CHEBI:29221
OCl(=O)(=O)=O
Safety and Handling

Safety and Handling

Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c

H271-H290-H302-H314-H335-H373

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-512-4

RTECSNumber : SC7500000

TSCA : Yes

Documents