missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric acid, 70%, 99.9985% (metals basis), Alfa Aesar™

Perchloric acid: 70%

Manufacturer:  Thermo Fisher Scientific 10983AK

 View more versions of this product

Catalog No. AA10983AK



Specifications

Specifications

Perchloric Acid 70%
250mL
Colorless liquid
ClHO4
1873
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
100.454
CHEBI:29221
Liquid
9 hPa (7mm Hg)
39°C (56mmHg)
White
7601-90-3
99.9985% (metals basis)
MFCD00011325
hclo4
OCl(=O)(=O)=O
1.67g/cm3 at 20°C
24247
100.46
Pungent
-112°C

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
hclo4
CHEBI:29221
OCl(=O)(=O)=O
Safety and Handling

Safety and Handling

H271-H314-H318-H302: May cause fire or explosion; strong oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: I

EINECSNumber : 231-512-4

Documents