missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric acid, 70%, 99.9985% (metals basis), Alfa Aesar™

Perchloric acid: 70%

Manufacturer:  Thermo Fisher Scientific 10983AD

 View more versions of this product

Catalog No. AA10983AD



Specifications

Specifications

Perchloric Acid 70%
50mL
Colorless liquid
ClHO4
1873
Total Metal Impurities: 0.0015% max. Concentration (weight/weight): 70% min
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
100.454
CHEBI:29221
Liquid
9 hPa (7mm Hg)
-112°C
White
7601-90-3
99.9985% (metals basis)
MFCD00011325
hclo4, hyperchloric acid, perchioric acid, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, ueberchlorsaeure, unii-v561v90bg2
Miscible with water.
OCl(=O)(=O)=O
1.67 g/cm3 at 20°C
24247
100.46
Pungent
113°C (235°F)
203°C

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
hclo4, hyperchloric acid, perchioric acid, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, ueberchlorsaeure, unii-v561v90bg2
CHEBI:29221
OCl(=O)(=O)=O
Safety and Handling

Safety and Handling

P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c

H271-H290-H302-H314-H335-H373

DOTInformation : DOT Class: 5.1, Packaging Group: I

EINECSNumber : 231-512-4

RTECSNumber : SC7500000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents