Please login to your online account to display your discounted pricing

Perchloric Acid, 0.7, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$510.64 - $1,583.66

Chemical Identifiers

Chemical Name or Material Perchloric Acid
CAS 7601-90-3
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula HClO4
Formula Weight 100.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003552

 
avantor performance
935905
Poly Bottle 2.5L Each for $1,583.66

02003553

 
avantor performance
935901
Poly Bottle 500mL Each for $510.64
Description & Specifications

Specifications

Formula Weight 100.46
Chemical Name or Material Perchloric Acid
Assay 70%
Molecular Formula HClO4
CAS 7601-90-3
Specific Gravity 1L = 1.68kg
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Packaging Poly Bottle
Quantity 2.5L

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use