Please login to your online account to display your discounted pricing

Pepstatin A, Fisher BioReagents


Quantity 100mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2671100

 View more versions of this product

Catalog No. BP2671100

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Pepstatin A
CAS 26305-03-3
ChemAlert Storage Symbol Gray
Identification Pass Test
Infrared Spectrum Conforms to reference
Recommended Storage RT
Packaging Amber Glass
Melting Range 230° to 240°C
Color White
Physical Form Powder/Solid
Melting Point 235°C
Molecular Formula C34H63N5O9
Quantity 100mg
Formula Weight 685.9
Merck Index 15, 7260
Synonym Pepstatin

Specifications

Chemical Name or Material Pepstatin A
CAS 26305-03-3
Physical Form Powder/Solid
Molecular Formula C34H63N5O9
Formula Weight 685.9
Synonym Pepstatin
Accessories
 

RT

SDS & Safety Documents